Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM7.1
Entrez Gene ID
417040
Gene Name
tripartite motif containing 7.1
Gene Aliases
TRIM7
Chromosome Location
Chr.16: 2482253 - 2492415 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099352.1 NP_001092822.1 6 - 7 1108 95
XM_015294950.2 XP_015150436.1 4 - 5 1197 95
XM_015294951.1 XP_015150437.1 5 - 6 1913 95
XM_015294952.1 XP_015150438.1 4 - 5 1546 95
XM_015294953.2 XP_015150439.1 4 - 5 966 95
XM_015294954.2 XP_015150440.1 4 - 5 1198 95
XM_015294955.1 XP_015150441.1 6 - 7 1885 95

Gene Symbol
LOC112533548
Entrez Gene ID
112533548
Gene Name
E3 ubiquitin-protein ligase TRIM7-like
Gene Aliases
-
Chromosome Location
Chr.16: 2376497 - 2384311 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025155849.1 XP_025011617.1 3 - 4 847 95

View Full Product Details