Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM7.2
Entrez Gene ID
425772
Gene Name
tripartite motif containing 7.2
Gene Aliases
TRIM27
Chromosome Location
Chr.16: 2454629 - 2458853 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099354.1 NP_001092824.1 5 - 6 954 68

View Full Product Details