Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPNVA
Entrez Gene ID
423974
Gene Name
vertebrate ancient opsin
Gene Aliases
CVAL, VA, VAL, VALOP
Chromosome Location
Chr.6: 36219396 - 36250696 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105318.1 NP_001098788.1 4 - 5 656 71
NM_001310089.1 NP_001297018.1 3 - 4 687 71
NM_001310090.1 NP_001297019.1 3 - 4 687 71
XM_015288991.2 XP_015144477.1 6 - 7 1569 71
XM_015288992.1 XP_015144478.1 3 - 4 856 71
GenBank mRNA AB295600.1 - 4 - 5 656 71
EF055883.2 - 3 - 4 687 71
GQ280390.1 - 3 - 4 687 71

View Full Product Details