Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGS2
Entrez Gene ID
378912
Gene Name
regulator of G-protein signaling 2
Gene Aliases
-
Chromosome Location
Chr.8: 3955419 - 3958244 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204395.2 NP_989726.1 1 - 2 155 71
GenBank mRNA AF502148.1 - 1 - 2 159 71
AF502149.1 - 1 - 2 213 71
BX931300.2 - 1 - 2 119 71
BX931885.1 - 1 - 2 113 71
CR386720.1 - 1 - 2 120 71

View Full Product Details