Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUT11
Entrez Gene ID
395071
Gene Name
fucosyltransferase 11
Gene Aliases
-
Chromosome Location
Chr.6: 16774824 - 16781124 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204428.1 NP_989759.1 1 - 2 775 61
GenBank mRNA AJ535752.1 - 1 - 2 775 61

View Full Product Details