Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR5A1
Entrez Gene ID
395960
Gene Name
nuclear receptor subfamily 5 group A member 1
Gene Aliases
Ad4BP, SF-1
Chromosome Location
Chr.17: 9728090 - 9744136 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205077.1 NP_990408.1 7 - 7 2337 105
GenBank mRNA AB002404.1 - 7 - 7 2337 105
CR353273.1 - 1 - 1 339 105

View Full Product Details