Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BFSP1
Entrez Gene ID
396056
Gene Name
beaded filament structural protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 12061525 - 12107620 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205151.1 NP_990482.1 3 - 4 760 98
XM_025148550.1 XP_025004318.1 10 - 11 3299 98
XM_025148551.1 XP_025004319.1 9 - 10 3189 98
GenBank mRNA X72873.1 - - 760 98

View Full Product Details