The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005825.1 | NP_001005825.1 | 7 - 8 | 909 | 137 |
XM_015292093.2 | XP_015147579.1 | 7 - 8 | 986 | 137 | |
GenBank mRNA | AJ720263.1 | - | 7 - 8 | 909 | 137 |