Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KPNA2
Entrez Gene ID
417420
Gene Name
karyopherin subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.18: 6978809 - 6986175 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006209.1 NP_001006209.1 9 - 10 1458 124
XM_015279891.2 XP_015135377.1 9 - 10 1495 124
GenBank mRNA AJ719475.1 - 9 - 10 1458 124

View Full Product Details