Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
427109
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr.Z: 58296404 - 58560287 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006574.1 NP_001006574.1 8 - 9 1018 87
XM_015280521.2 XP_015136007.1 7 - 8 856 87
GenBank mRNA AJ720240.1 - 8 - 9 1018 87

View Full Product Details