Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDC4
Entrez Gene ID
419184
Gene Name
syndecan 4
Gene Aliases
syndecan-4
Chromosome Location
Chr.20: 5127360 - 5141905 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007869.1 NP_001007870.1 4 - 5 437 77

View Full Product Details