Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LZIC
Entrez Gene ID
419445
Gene Name
leucine zipper and CTNNBIP1 domain containing
Gene Aliases
-
Chromosome Location
Chr.21: 3578448 - 3601156 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008450.1 NP_001008450.1 2 - 3 163 122
XM_015296891.2 XP_015152377.1 2 - 3 359 122
XM_015296892.2 XP_015152378.1 2 - 3 289 122
XM_025142520.1 XP_024998288.1 2 - 3 196 122
XM_025142521.1 XP_024998289.1 2 - 3 336 122
GenBank mRNA AJ719972.1 - 2 - 3 163 122

View Full Product Details