Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA5
Entrez Gene ID
386577
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
-
Chromosome Location
Chr.10: 4080158 - 4096756 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204415.2 NP_989746.2 3 - 4 313 83
XM_025153788.1 XP_025009556.1 3 - 4 375 83
XM_025153789.1 XP_025009557.1 4 - 5 1158 83
XM_025153790.1 XP_025009558.1 3 - 4 1471 83
XM_025153791.1 XP_025009559.1 3 - 4 320 83
GenBank mRNA J05642.1 - 3 - 4 258 83

View Full Product Details