Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRD4
Entrez Gene ID
423016
Gene Name
dopamine receptor D4
Gene Aliases
D4
Chromosome Location
Chr.5: 785918 - 791782 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142849.2 NP_001136321.1 2 - 3 438 78

View Full Product Details