Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
OPN1SW
Entrez Gene ID
396419
Gene Name
opsin 1 (cone pigments), short-wave-sensitive (violet cone opsin)
Gene Aliases
SWS1, opsin
Chromosome Location
Chr.1: 752193 - 752615 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205438.1 NP_990769.1 - 1251 117
GenBank mRNA M92039.1 - - 1251 117

Gene Symbol
AGAP1
Entrez Gene ID
424024
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
CENTG2, CENTG3
Chromosome Location
Chr.7: 5226664 - 5522474 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942493.3 XP_004942550.1 17 - 17 7072 117
XM_015289260.2 XP_015144746.1 20 - 20 8650 117
XM_015289261.2 XP_015144747.1 19 - 19 8637 117
XM_015289263.2 XP_015144749.1 19 - 19 8570 117
XM_015289264.2 XP_015144750.1 19 - 19 8496 117
XM_015289265.2 XP_015144751.1 19 - 19 8490 117
XM_015289267.2 XP_015144753.1 20 - 20 7583 117
XM_015289268.2 XP_015144754.1 19 - 19 7312 117
XM_421880.6 XP_421880.3 18 - 18 7234 117

View Full Product Details