Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLU
Entrez Gene ID
548016
Gene Name
ferredoxin-dependent glutamate synthase
Gene Aliases
-
Chromosome Location
Chr.3: 36294653 - 36322791 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF039851.1 - 11 - 12 1334 100
AK287031.1 - 20 - 21 3158 100

View Full Product Details