Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYS
Entrez Gene ID
548023
Gene Name
cystathionine-gamma-synthase precursor
Gene Aliases
-
Chromosome Location
Chr.9: 42595767 - 42600602 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250643.2 NP_001237572.1 6 - 7 1160 93
GenBank mRNA AF141602.1 - 6 - 7 1165 93
AK244280.1 - 6 - 7 1183 93
AK286711.1 - 6 - 7 1190 93

View Full Product Details