Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CHLH
- Entrez Gene ID
-
548043
- Gene Name
- magnesium chelatase subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 37317303 - 37323760 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100787735
- Entrez Gene ID
-
100787735
- Gene Name
- magnesium-chelatase subunit ChlH, chloroplastic-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 39842978 - 39849601 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK286404.1 |
- |
3 - 4 |
3468 |
69 |
View Full Product Details