Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUT1
Entrez Gene ID
547679
Gene Name
sucrose transporter
Gene Aliases
-
Chromosome Location
Chr.10: 44376307 - 44379793 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249369.1 NP_001236298.1 2 - 3 1357 102
GenBank mRNA AJ563364.1 - 2 - 3 1357 102
AK244954.1 - 2 - 3 1403 102

View Full Product Details