Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP3
Entrez Gene ID
547708
Gene Name
purple acid phosphatase-like protein
Gene Aliases
phytase
Chromosome Location
Chr.17: 8838656 - 8843496 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249748.1 NP_001236677.1 3 - 4 482 89
GenBank mRNA AK244377.1 - 3 - 4 513 89
AK286765.1 - 3 - 4 539 89
AY151271.1 - 3 - 4 482 89
AY151272.1 - 3 - 4 482 89
AY151273.1 - 3 - 4 482 89
AY151274.1 - 3 - 4 482 89
AY151275.1 - 3 - 4 482 89
GQ422778.1 - 3 - 4 482 89

View Full Product Details