Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAP3
Entrez Gene ID
547708
Gene Name
purple acid phosphatase-like protein
Gene Aliases
phytase
Chromosome Location
Chr.17: 8838656 - 8843496 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249748.1 NP_001236677.1 5 - 6 826 99
GenBank mRNA AK244377.1 - 5 - 6 857 99
AK286765.1 - 5 - 6 883 99
AY151271.1 - 5 - 6 826 99
AY151272.1 - 5 - 6 826 99
AY151273.1 - 5 - 6 826 99
AY151274.1 - 5 - 6 826 99
AY151275.1 - 5 - 6 826 99
GQ422778.1 - 5 - 6 826 99

View Full Product Details