Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPA1
Entrez Gene ID
547804
Gene Name
G protein alpha subunit
Gene Aliases
-
Chromosome Location
Chr.4: 4544027 - 4549811 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251255.1 NP_001238184.1 2 - 3 198 84
GenBank mRNA L27418.1 - 2 - 3 198 84

View Full Product Details