Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCCA
Entrez Gene ID
548094
Gene Name
3-methylcrotonyl-CoA carboxylase, biotin-carrier domain
Gene Aliases
-
Chromosome Location
Chr.2: 14806148 - 14812185 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA U08469.1 - 3 - 4 417 94

View Full Product Details