Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COI1
Entrez Gene ID
732662
Gene Name
coronatine-insensitive 1
Gene Aliases
-
Chromosome Location
Chr.11: 36678304 - 36682125 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251661.1 NP_001238590.1 1 - 2 478 88
GenBank mRNA DQ136026.1 - 1 - 2 478 88

View Full Product Details