Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY57
Entrez Gene ID
100170682
Gene Name
transcription factor
Gene Aliases
-
Chromosome Location
Chr.18: 54287790 - 54289766 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250690.2 NP_001237619.1 1 - 2 376 76
GenBank mRNA BT098858.1 - 1 - 2 391 76
EU375353.1 - 1 - 2 377 76

View Full Product Details