Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZIP61
Entrez Gene ID
780555
Gene Name
bZIP transcription factor bZIP61
Gene Aliases
-
Chromosome Location
Chr.4: 6479342 - 6485087 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BT098351.1 - 1 - 1 172 99
DQ787019.1 - 1 - 1 45 99

View Full Product Details