Gene Transcripts

Gene Symbol
ALDH7A1
Entrez Gene ID
501
Gene Name
aldehyde dehydrogenase 7 family member A1
Gene Aliases
ATQ1, EPD, PDE
Chromosome Location
Chr. 5: 126541841 - 126595390 on Build GRCh38
UniGene ID
Hs.483239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001182.4 NP_001173.2 5, 6 700
NM_001201377.1 NP_001188306.1 5, 6 700
NM_001202404.1 NP_001189333.2 5, 6 700
XM_011543417.2 XP_011541719.1 5, 6 691
XM_017009493.1 XP_016864982.1 5, 6 476
GenBank mRNA AK297365.1 - 5, 6 646
AK312459.1 - 5, 6 529
AU135961.1 - 5, 6 558
BC002515.2 - 5, 6 517 3139235
BC071712.1 - 5, 6 494 5518212
BC073174.1 - 5, 6 486 6501646
S74728.1 - 5, 6 478

View Full Product Details