Gene Transcripts
- Gene Symbol
- CBS
- Entrez Gene ID
- 875
- Gene Name
- cystathionine-beta-synthase
- Gene Aliases
- HIP4
- Chromosome Location
- Chr. 21: 43053190 - 43076868 on Build GRCh38
- UniGene ID
- Hs.533013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000071.2 |
NP_000062.1
|
9, 10 |
1063 |
|
NM_001178008.2 |
NP_001171479.1
|
9, 10 |
1070 |
|
NM_001178009.2 |
NP_001171480.1
|
9, 10 |
1070 |
|
NM_001320298.1 |
NP_001307227.1
|
10, 11 |
1064 |
|
NM_001321072.1 |
NP_001308001.1
|
6, 7 |
1129 |
|
XM_011529773.2 |
XP_011528075.1
|
7, 8 |
1189 |
|
XM_011529774.2 |
XP_011528076.1
|
7, 8 |
1189 |
|
XM_011529775.2 |
XP_011528077.1
|
7, 8 |
1189 |
|
XM_011529776.2 |
XP_011528078.1
|
7, 8 |
1189 |
|
XM_011529777.1 |
XP_011528079.1
|
9, 10 |
1006 |
|
XM_011529781.1 |
XP_011528083.1
|
10, 11 |
1071 |
|
XM_011529782.1 |
XP_011528084.1
|
9, 10 |
1469 |
|
XM_011529783.2 |
XP_011528085.1
|
6, 7 |
1101 |
|
XM_017028489.1 |
XP_016883978.1
|
10, 11 |
1466 |
|
XM_017028490.1 |
XP_016883979.1
|
10, 11 |
1463 |
|
XM_017028491.1 |
XP_016883980.1
|
9, 10 |
1006 |
|
XM_017028492.1 |
XP_016883981.1
|
10, 11 |
1071 |
|
XM_017028493.1 |
XP_016883982.1
|
6, 7 |
1300 |
|
GenBank mRNA |
AK294608.1 |
- |
11, 12 |
935 |
|
AK301063.1 |
- |
7, 8 |
665 |
|
AK313691.1 |
- |
9, 10 |
968 |
|
AL137314.1 |
- |
6, 7 |
590 |
|
BC000440.2 |
- |
9, 10 |
938 |
2820694 |
BC010242.1 |
- |
9, 10 |
987 |
4154798 |
BC011381.1 |
- |
9, 10 |
926 |
3869615 |
BT007154.1 |
- |
7, 8 |
818 |
|
L00972.1 |
- |
6, 7 |
1141 |
|
L14577.1 |
- |
9, 10 |
976 |
|
L19501.1 |
- |
9, 10 |
999 |
|
X82166.1 |
- |
9, 10 |
965 |
|
- Gene Symbol
- CBSL
- Entrez Gene ID
- 102724560
- Gene Name
- cystathionine-beta-synthase like
- Gene Aliases
- Chromosome Location
- Chr. 21: 6444868 - 6468606 on Build GRCh38
- UniGene ID
- Hs.533013
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321073.1 |
NP_001308002.1
|
9, 10 |
1070 |
|
XM_011546094.1 |
XP_011544396.1
|
9, 10 |
962 |
|
XM_011546095.2 |
XP_011544397.1
|
9, 10 |
1104 |
|
XM_011546097.2 |
XP_011544399.1
|
10, 11 |
1131 |
|
XM_011546098.1 |
XP_011544400.1
|
9, 10 |
1469 |
|
XM_011546099.1 |
XP_011544401.1
|
9, 10 |
962 |
|
XM_017028209.1 |
XP_016883698.1
|
7, 8 |
1167 |
|
XM_017028210.1 |
XP_016883699.1
|
7, 8 |
1167 |
|
XM_017028211.1 |
XP_016883700.1
|
10, 11 |
1467 |
|
XM_017028212.1 |
XP_016883701.1
|
11, 12 |
1523 |
|
XM_017028213.1 |
XP_016883702.1
|
9, 10 |
962 |
|
XM_017028214.1 |
XP_016883703.1
|
10, 11 |
1464 |
|
XM_017028215.1 |
XP_016883704.1
|
10, 11 |
1079 |
|
XM_017028216.1 |
XP_016883705.1
|
9, 10 |
1469 |
|
XM_017028217.1 |
XP_016883706.1
|
9, 10 |
962 |
|
XM_017028218.1 |
XP_016883707.1
|
10, 11 |
1131 |
|
GenBank mRNA |
BC007257.1 |
- |
9, 10 |
1040 |
|
View Full Product Details