Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004060.3 | NP_004051.1 | 2 | 359 | |
NM_199246.1 | NP_954854.1 | 3 | 259 | ||
GenBank mRNA | AK312913.1 | - | 2 | 335 | |
BC000196.1 | - | 3 | 209 | 3351902 | |
BC007093.2 | - | 3 | 234 | 4251051 | |
BM479568.1 | - | 3 | 272 | ||
BT007134.1 | - | 1 | 125 | ||
BX644635.1 | - | 2 | 350 | ||
D78341.1 | - | 2 | 315 | ||
L49504.1 | - | 2 | 336 | ||
U47413.1 | - | 2 | 338 | ||
U53328.1 | - | 1 | 138 | ||
X77794.1 | - | 1 | 107 |