Gene Transcripts
- Gene Symbol
- CDH12
- Entrez Gene ID
- 1010
- Gene Name
- cadherin 12
- Gene Aliases
- CDHB
- Chromosome Location
- Chr. 5: 21750191 - 22853622 on Build GRCh38
- UniGene ID
- Hs.113684
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317227.1 |
NP_001304156.1
|
8, 9 |
1849 |
|
NM_001317228.1 |
NP_001304157.1
|
8, 9 |
1844 |
|
NM_004061.4 |
NP_004052.2
|
11, 12 |
2478 |
|
XM_011513927.2 |
XP_011512229.1
|
9, 10 |
1755 |
|
XM_017008920.1 |
XP_016864409.1
|
10, 11 |
2105 |
|
XM_017008921.1 |
XP_016864410.1
|
11, 12 |
2198 |
|
GenBank mRNA |
AK092176.1 |
- |
6, 7 |
1128 |
|
AK295123.1 |
- |
8, 9 |
1844 |
|
AK314800.1 |
- |
8, 9 |
1849 |
|
BC047608.1 |
- |
11, 12 |
2183 |
5751948 |
L33477.1 |
- |
11, 12 |
2479 |
|
L34057.1 |
- |
7, 8 |
1484 |
|
View Full Product Details