Gene Transcripts
- Gene Symbol
- DCT
- Entrez Gene ID
- 1638
- Gene Name
- dopachrome tautomerase
- Gene Aliases
- TRP-2, TYRP2
- Chromosome Location
- Chr. 13: 94437304 - 94550265 on Build GRCh38
- UniGene ID
- Hs.301865
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001129889.2 |
NP_001123361.1
|
5, 6 |
1480 |
|
NM_001322182.1 |
NP_001309111.1
|
4, 5 |
702 |
|
NM_001322183.1 |
NP_001309112.1
|
5, 6 |
756 |
|
NM_001322184.1 |
NP_001309113.1
|
6, 7 |
1056 |
|
NM_001322185.1 |
NP_001309114.1
|
5, 6 |
1002 |
|
NM_001322186.1 |
NP_001309115.1
|
7, 8 |
1271 |
|
NM_001922.4 |
NP_001913.2
|
5, 6 |
1480 |
|
XM_011521049.1 |
XP_011519351.1
|
7, 8 |
2104 |
|
XM_017020401.1 |
XP_016875890.1
|
5, 6 |
1483 |
|
GenBank mRNA |
AK293115.1 |
- |
5, 6 |
1263 |
|
BC028311.1 |
- |
5, 6 |
1327 |
4705935 |
D17547.1 |
- |
5, 6 |
1467 |
|
DQ902581.1 |
- |
5, 6 |
1345 |
|
L18967.1 |
- |
5, 6 |
1437 |
|
S69231.1 |
- |
5, 6 |
1402 |
|
View Full Product Details