Gene Transcripts

Gene Symbol
DPT
Entrez Gene ID
1805
Gene Name
dermatopontin
Gene Aliases
TRAMP
Chromosome Location
Chr. 1: 168695457 - 168729204 on Build GRCh38
UniGene ID
Hs.80552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001937.4 NP_001928.2 2 439
GenBank mRNA BC033736.1 - 2 425
BP257325.1 - 2 439
Z22865.1 - 2 421

View Full Product Details