Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001406.3 | NP_001397.1 | 2 | 790 | |
GenBank mRNA | AK314841.1 | - | 2 | 896 | |
BC022499.1 | - | 1 | 229 | ||
BC042944.2 | - | 2 | 790 | 5286243 | |
U57001.1 | - | 2 | 790 | ||
U62775.1 | - | 2 | 594 | ||
U66406.1 | - | 2 | 1006 |