Gene Transcripts

Gene Symbol
FDFT1
Entrez Gene ID
2222
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
DGPT, ERG9, SQS, SS
Chromosome Location
Chr. 8: 11795573 - 11839309 on Build GRCh38
UniGene ID
Hs.593928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287742.1 NP_001274671.1 4 870
NM_001287743.1 NP_001274672.1 3 672
NM_001287744.1 NP_001274673.1 2 499
NM_001287745.1 NP_001274674.1 3 578
NM_001287747.1 NP_001274676.1 2 535
NM_001287748.1 NP_001274677.1 2 197
NM_001287749.1 NP_001274678.1 2 435
NM_001287750.1 NP_001274679.1 1 437
NM_004462.4 NP_004453.3 2 381
XM_017013217.1 XP_016868706.1 4 1162
GenBank mRNA AK057726.1 - 2 499
AK098682.1 - 955
AK293545.1 - 2 435
AK296043.1 - 2 253
AK297868.1 - 1 380
AK311362.1 - 2 567
AK315993.1 - 3 448
AK316033.1 - 2 402
AK316182.1 - 1 397
AK316351.1 - 2 162
AK316531.1 - 2 147
BC003573.1 - 2 228 3538137
BC009251.2 - 2 228 2961623
BC029641.1 - 2 191 4514761
BC034440.1 - 2 191
BE384903.1 - 2 535
BG752446.1 - 245
BI765996.1 - 4 474
BT006704.1 - 2 159
CR457033.1 - 2 159
DC314061.1 - 2 357
DC387127.1 - 1 431
DC387479.1 - 2 197
DC396859.1 - 2 383
L06070.1 - 2 168
L06105.1 - 2 203
S76822.1 - 2 224
X69141.1 - 2 250

View Full Product Details