Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005911.5 | NP_005902.1 | 2 | 425 | |
GenBank mRNA | AK291126.1 | - | 2 | 233 | |
AK293841.1 | - | 2 | 235 | ||
AK297758.1 | - | 2 | 128 | ||
AK308591.1 | - | 2 | 128 | ||
AK308758.1 | - | 2 | 93 | ||
AK309338.1 | - | 2 | 233 | ||
AK316411.1 | - | 2 | 383 | ||
BC001686.2 | - | 2 | 222 | 3010820 | |
BC001854.2 | - | 2 | 222 | 3010820 | |
BI549760.1 | - | 2 | 244 | ||
DC322518.1 | - | 2 | 266 | ||
DQ083239.1 | - | 2 | 161 | ||
X68836.1 | - | 2 | 180 |