Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005370.4 | NP_005361.2 | 1 | 236 | |
XM_011528020.2 | XP_011526322.1 | 1 | 24 | ||
GenBank mRNA | AF498943.1 | - | 1 | 14 | |
AK025165.1 | - | 1 | 259 | ||
AK293676.1 | - | 1 | 19 | ||
BC002977.1 | - | 1 | 90 | 3547214 | |
BT007184.1 | - | 1 | 14 | ||
CR536583.1 | - | 1 | 14 | ||
CR542274.1 | - | 1 | 14 | ||
X56741.1 | - | 1 | 24 |