Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005399.4 | NP_005390.1 | 5 | 617 | |
NR_103871.1 | 4 | 450 | |||
XM_011509729.1 | XP_011508031.1 | 5 | 617 | ||
GenBank mRNA | AJ224538.1 | - | 4 | 433 | |
AK292820.1 | - | 5 | 498 | ||
AK294863.1 | - | 4 | 422 | ||
AK310982.1 | - | 5 | 498 | ||
AK316005.1 | - | 4 | 328 | ||
AL037763.1 | - | 4 | 414 | ||
BC053610.1 | - | 5 | 572 | 6091476 | |
KJ534919.1 | - | 4 | 433 | ||
KJ535024.1 | - | 3 | 266 | ||
KJ535025.1 | - | 4 | 433 |