Gene Transcripts

Gene Symbol
PSMC6
Entrez Gene ID
5706
Gene Name
proteasome 26S subunit, ATPase 6
Gene Aliases
CADP44, HEL-S-73, P44, SUG2, p42
Chromosome Location
Chr. 14: 52707163 - 52727998 on Build GRCh38
UniGene ID
Hs.156171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002806.3 NP_002797.3 8 614
XM_017021472.1 XP_016876961.1 8 2117
GenBank mRNA AF006305.1 - 8 587
AK299155.1 - 7 528
AK313670.1 - 8 582
BC005390.1 - 8 592 3996906
BT006843.1 - 8 572
CR456709.1 - 8 572
D78275.1 - 8 588
FJ224299.1 - 8 588

View Full Product Details