Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005061.2 | NP_005052.1 | 8 | 1040 | |
XM_011522571.2 | XP_011520873.1 | 8 | 1500 | ||
GenBank mRNA | AK313162.1 | - | 8 | 1052 | |
BC050413.1 | - | 8 | 1032 | 6204847 | |
U65581.1 | - | 8 | 1041 |