Gene Transcripts

Gene Symbol
UBE3A
Entrez Gene ID
7337
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location
Chr. 15: 25337234 - 25439086 on Build GRCh38
UniGene ID
Hs.598862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000462.3 NP_000453.2 7 1492
NM_130838.1 NP_570853.1 3 707
NM_130839.2 NP_570854.1 6 1427
XM_005268267.4 XP_005268324.1 8 1794
XM_005268268.4 XP_005268325.1 7 1681
XM_005268269.4 XP_005268326.1 8 1525
XM_005268270.4 XP_005268327.1 7 1482
XM_005268271.4 XP_005268328.1 4 1301
XM_006720675.3 XP_006720738.1 8 1746
XM_006720676.3 XP_006720739.1 5 991
XM_011521994.2 XP_011520296.1 8 1683
XM_011521995.2 XP_011520297.1 8 1357
XM_017022544.1 XP_016878033.1 9 1719
XM_017022545.1 XP_016878034.1 8 1678
XM_017022546.1 XP_016878035.1 8 1586
XM_017022547.1 XP_016878036.1 7 1618
XM_017022548.1 XP_016878037.1 7 1613
XM_017022549.1 XP_016878038.1 7 1420
XM_017022550.1 XP_016878039.1 7 1572
XM_017022551.1 XP_016878040.1 8 1623
XM_017022552.1 XP_016878041.1 8 1313
XM_017022553.1 XP_016878042.1 8 1628
XM_017022554.1 XP_016878043.1 4 1632
XM_017022555.1 XP_016878044.1 6 1401
XM_017022556.1 XP_016878045.1 7 1550
XM_017022557.1 XP_016878046.1 6 1485
XM_017022558.1 XP_016878047.1 7 1681
XM_017022559.1 XP_016878048.1 4 1301
GenBank mRNA AF273050.1 - 4 762
AK056239.1 - 6 1395
AK130491.1 - 2 436
AK291405.1 - 4 853
AK292514.1 - 6 1037
BC002582.2 - 6 1380 3160225
L07557.1 - 3 702
U84404.1 - 8 1249
X98031.1 - 4 732
X98032.1 - 2 663
X98033.1 - 4 723

View Full Product Details