Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256910.1 | NP_001243839.1 | 12 | 2107 | |
NM_004728.3 | NP_004719.2 | 12 | 1988 | ||
XM_011540336.1 | XP_011538638.1 | 11 | 1818 | ||
XM_017016910.1 | XP_016872399.1 | 11 | 1736 | ||
GenBank mRNA | AB062394.1 | - | 4 | 467 | |
AK025629.1 | - | 9 | 1290 | ||
AK315585.1 | - | 12 | 1937 | ||
BC008071.2 | - | 12 | 1926 | 2987980 | |
BX648405.1 | - | 12 | 1874 | ||
CR749598.1 | - | 12 | 2108 | ||
U41387.1 | - | 12 | 1939 |