Gene Transcripts
- Gene Symbol
- TRIP13
- Entrez Gene ID
- 9319
- Gene Name
- thyroid hormone receptor interactor 13
- Gene Aliases
- 16E1BP
- Chromosome Location
- Chr. 5: 892854 - 919348 on Build GRCh38
- UniGene ID
- Hs.436187
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166260.1 |
NP_001159732.1
|
2, 3 |
413 |
|
NM_004237.3 |
NP_004228.1
|
2, 3 |
413 |
|
XM_011514163.2 |
XP_011512465.1
|
2, 3 |
383 |
|
GenBank mRNA |
AK310376.1 |
- |
2, 3 |
383 |
|
BC000404.2 |
- |
2, 3 |
378 |
2822981 |
BC019294.1 |
- |
2, 3 |
378 |
2822981 |
BG722761.1 |
- |
2, 3 |
415 |
|
CR456744.1 |
- |
2, 3 |
268 |
|
L40384.1 |
- |
2, 3 |
274 |
|
U96131.1 |
- |
2, 3 |
313 |
|
View Full Product Details