Gene Transcripts

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
Chromosome Location
Chr. 14: 24171853 - 24180408 on Build GRCh38
UniGene ID
Hs.419259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048205.1 NP_001041670.1 14 1710
NM_005132.2 NP_005123.2 15 1609
XM_011537421.1 XP_011535723.1 14 1882
XM_017021841.1 XP_016877330.1 14 1756
GenBank mRNA AF006264.1 - 14 1182
AK001435.1 - 15 1609
AK291191.1 - 15 1741
AL832973.1 - 12 2099
BC004159.2 - 14 1266 2822680
BC010887.1 - 14 1643 3535425
BC019326.1 - 8 628
CR457263.1 - 14 1182

View Full Product Details