Gene Transcripts

Gene Symbol
SACM1L
Entrez Gene ID
22908
Gene Name
SAC1 suppressor of actin mutations 1-like (yeast)
Gene Aliases
SAC1
Chromosome Location
Chr. 3: 45689241 - 45745425 on Build GRCh38
UniGene ID
Hs.156509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319072.1 NP_001306001.1 15, 16 1486
NM_001319073.1 NP_001306002.1 17, 18 1654
NM_014016.4 NP_054735.3 16, 17 1614
XM_011533500.2 XP_011531802.1 16, 17 1405
GenBank mRNA AB020658.1 - 16, 17 1458
AK126434.1 - 3, 4 929
AK291142.1 - 16, 17 1498
AK296421.1 - 15, 16 1268
AK301248.1 - 17, 18 1494
AL136831.1 - 16, 17 1404
BC016559.1 - 16, 17 1410 3049075
CR749564.1 - 16, 17 2550

View Full Product Details