Gene Transcripts

Gene Symbol
NIPSNAP3A
Entrez Gene ID
25934
Gene Name
nipsnap homolog 3A
Gene Aliases
HSPC299, NIPSNAP4, TASSC
Chromosome Location
Chr. 9: 104747688 - 104760122 on Build GRCh38
UniGene ID
Hs.530275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015469.1 NP_056284.1 4 666
GenBank mRNA AF161417.1 - 4 647
AK024015.1 - 4 627
AK301410.1 - 4 657
AL117557.1 - 4 610
BC005935.1 - 4 666 4064889
BC045830.1 - 4 606

View Full Product Details