Gene Transcripts

Gene Symbol
FBXL5
Entrez Gene ID
26234
Gene Name
F-box and leucine rich repeat protein 5
Gene Aliases
FBL4, FBL5, FLR1
Chromosome Location
Chr. 4: 15604384 - 15681545 on Build GRCh38
UniGene ID
Hs.643433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193534.1 NP_001180463.1 9 1757
NM_001193535.1 NP_001180464.1 9 1709
NM_012161.3 NP_036293.1 9 1760
NR_036464.1 10 1864
XM_006713959.3 XP_006714022.1 10 1856
XM_011513831.2 XP_011512133.1 9 1542
XM_011513832.2 XP_011512134.1 9 1689
XM_011513833.2 XP_011512135.1 8 1500
XM_017008019.1 XP_016863508.1 11 2627
GenBank mRNA AF052097.1 - 1 319
AF142481.1 - 9 1682
AF157323.1 - 9 1621
AF174591.1 - 6 1167
AF176700.1 - 9 1644
AF199420.1 - 9 1646
AK000153.1 - 8 1470
AK295510.1 - 9 1679
AK310860.1 - 9 1689
AL832808.1 - 7 1396
BC012320.1 - 10 1793
BC030656.1 - 9 1731 4837080
BX537957.1 - 9 1655

View Full Product Details