Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014384.2 | NP_055199.1 | 11 | 1275 | |
XM_005271505.3 | XP_005271562.1 | 11 | 2298 | ||
XM_017017542.1 | XP_016873031.1 | 12 | 1503 | ||
XM_017017543.1 | XP_016873032.1 | 12 | 2069 | ||
XM_017017544.1 | XP_016873033.1 | 12 | 1428 | ||
XM_017017545.1 | XP_016873034.1 | 11 | 2604 | ||
XM_017017546.1 | XP_016873035.1 | 11 | 1328 | ||
XM_017017547.1 | XP_016873036.1 | 10 | 1108 | ||
XM_017017549.1 | XP_016873038.1 | 12 | 2443 | ||
GenBank mRNA | AB209538.1 | - | 10 | 2392 | |
AF126245.1 | - | 11 | 1302 | ||
AK000359.1 | - | 11 | 1247 | ||
AK026243.1 | - | 3 | 266 | ||
AK074640.1 | - | 8 | 874 | ||
AK124235.1 | - | 1 | 1571 | ||
AK125373.1 | - | 8 | 2273 | ||
BC001964.1 | - | 11 | 1244 | 3462406 |