Gene Transcripts
- Gene Symbol
- POMT2
- Entrez Gene ID
- 29954
- Gene Name
- protein O-mannosyltransferase 2
- Gene Aliases
- LGMD2N, MDDGA2, MDDGB2, MDDGC2
- Chromosome Location
- Chr. 14: 77274956 - 77320885 on Build GRCh38
- UniGene ID
- Hs.132989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013382.5 |
NP_037514.2
|
2, 3 |
534 |
|
XM_011536675.2 |
XP_011534977.1
|
2, 3 |
534 |
|
XM_011536677.2 |
XP_011534979.1
|
2, 3 |
535 |
|
XM_017021244.1 |
XP_016876733.1
|
2, 3 |
528 |
|
GenBank mRNA |
AB209114.1 |
- |
2, 3 |
356 |
|
AF105020.1 |
- |
2, 3 |
537 |
|
AL554279.3 |
- |
2, 3 |
398 |
|
AY090480.1 |
- |
2, 3 |
537 |
|
BC031651.1 |
- |
2, 3 |
424 |
5169145 |
CR627447.1 |
- |
2, 3 |
104 |
|
View Full Product Details