Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198976.2 | NP_945327.2 | 14 | 1762 | |
GenBank mRNA | AF161479.1 | - | 14 | 1692 | |
AJ238374.1 | - | 8 | 1504 | ||
AJ238375.1 | - | 9 | 1085 | ||
AJ238379.1 | - | 14 | 1696 | ||
AK001316.1 | - | 14 | 1670 | ||
AK023927.1 | - | 14 | 1687 | ||
AK127431.1 | - | 3 | 3039 | ||
AK293410.1 | - | 14 | 1714 | ||
BC014952.1 | - | 14 | 1698 | 4860894 | |
BM974193.1 | - | 2 | 556 | ||
BX647417.1 | - | 14 | 1697 |