Gene Transcripts

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
Chromosome Location
Chr. 2: 233681938 - 233773299 on Build GRCh38
UniGene ID
Hs.554822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019077.2 NP_061950.2 1 304
GenBank mRNA AY435142.1 - 1 304
DQ383513.1 - 1 16
U89507.1 - 1 304

View Full Product Details